Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCTCAGATGAAGGTGGTGGTT[G/T]TGATGGAGTTGGGTGTGAGGATAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611337 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB8 - DnaJ heat shock protein family (Hsp40) member B8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153330.4 | 200 | UTR 5 | NP_699161.1 | |||
XM_006713519.3 | 200 | Intron | XP_006713582.1 |
DNAJB8-AS1 - DNAJB8 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |