Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607342 MIM: 180472 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPEB1 - cytoplasmic polyadenylation element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079533.1 | Intron | NP_001073001.1 | ||||
NM_001079534.1 | Intron | NP_001073002.1 | ||||
NM_001079535.1 | Intron | NP_001073003.1 | ||||
NM_001288819.1 | Intron | NP_001275748.1 | ||||
NM_001288820.1 | Intron | NP_001275749.1 | ||||
NM_030594.4 | Intron | NP_085097.3 |
RPS17 - ribosomal protein S17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |