Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610660 MIM: 614574 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GLYR1 - glyoxylate reductase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308096.1 | 3248 | Intron | NP_001295025.1 | |||
NM_001324096.1 | 3248 | Intron | NP_001311025.1 | |||
NM_001324097.1 | 3248 | Intron | NP_001311026.1 | |||
NM_001324098.1 | 3248 | Intron | NP_001311027.1 | |||
NM_032569.3 | 3248 | Intron | NP_115958.2 | |||
XM_005255639.4 | 3248 | Intron | XP_005255696.2 | |||
XM_011522716.2 | 3248 | UTR 3 | XP_011521018.1 | |||
XM_011522717.1 | 3248 | Intron | XP_011521019.1 | |||
XM_017023786.1 | 3248 | Intron | XP_016879275.1 | |||
XM_017023787.1 | 3248 | Intron | XP_016879276.1 | |||
XM_017023788.1 | 3248 | Intron | XP_016879277.1 | |||
XM_017023789.1 | 3248 | Intron | XP_016879278.1 |
ROGDI - rogdi homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024589.2 | 3248 | Intron | NP_078865.1 | |||
XM_006720947.3 | 3248 | Intron | XP_006721010.1 | |||
XM_006720948.3 | 3248 | Intron | XP_006721011.1 |
SMIM22 - small integral membrane protein 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |