Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614677 MIM: 603892 MIM: 137780 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC103 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC103 - coiled-coil domain containing 103 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258395.1 | 113 | UTR 5 | NP_001245324.1 | |||
NM_001258396.1 | 113 | UTR 5 | NP_001245325.1 | |||
NM_001258397.1 | 113 | UTR 5 | NP_001245326.1 | |||
NM_001258398.1 | 113 | UTR 5 | NP_001245327.1 | |||
NM_001258399.1 | 113 | UTR 5 | NP_001245328.1 | |||
NM_213607.2 | 113 | UTR 5 | NP_998772.1 |
EFTUD2 - elongation factor Tu GTP binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142605.1 | 113 | Intron | NP_001136077.1 | |||
NM_001258353.1 | 113 | Intron | NP_001245282.1 | |||
NM_001258354.1 | 113 | Intron | NP_001245283.1 | |||
NM_004247.3 | 113 | Intron | NP_004238.3 |
GFAP - glial fibrillary acidic protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |