Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609742 MIM: 616659 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL4I1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258017.1 | 2130 | Missense Mutation | GCC,TCC | A,S 523 | NP_001244946.1 | |
NM_001258018.1 | 2130 | Missense Mutation | GCC,TCC | A,S 523 | NP_001244947.1 | |
NM_152899.1 | 2130 | Missense Mutation | GCC,TCC | A,S 501 | NP_690863.1 | |
NM_172374.2 | 2130 | Missense Mutation | GCC,TCC | A,S 523 | NP_758962.1 |
MIR4750 - microRNA 4750 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D17 - TBC1 domain family member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |