Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616582 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LSR - lipolysis stimulated lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260489.1 | 715 | Silent Mutation | GTC,GTT | V,V 164 | NP_001247418.1 | |
NM_001260490.1 | 715 | Silent Mutation | GTC,GTT | V,V 164 | NP_001247419.1 | |
NM_015925.6 | 715 | Silent Mutation | GTC,GTT | V,V 164 | NP_057009.3 | |
NM_205834.3 | 715 | Silent Mutation | GTC,GTT | V,V 164 | NP_991403.1 | |
NM_205835.3 | 715 | Silent Mutation | GTC,GTT | V,V 164 | NP_991404.1 | |
XM_005258980.1 | 715 | Silent Mutation | GTC,GTT | V,V 164 | XP_005259037.1 | |
XM_005258982.1 | 715 | Silent Mutation | GTC,GTT | V,V 164 | XP_005259039.1 | |
XM_011527026.1 | 715 | Silent Mutation | GTC,GTT | V,V 164 | XP_011525328.1 |
Set Membership: |
HapMap Validated |