Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614612 | ||||||||||||||||||||
Literature Links: |
KANK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KANK4 - KN motif and ankyrin repeat domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320269.1 | Intron | NP_001307198.1 | ||||
NM_181712.4 | Intron | NP_859063.3 | ||||
XM_011540843.2 | Intron | XP_011539145.1 | ||||
XM_011540845.2 | Intron | XP_011539147.1 | ||||
XM_017000483.1 | Intron | XP_016855972.1 | ||||
XM_017000484.1 | Intron | XP_016855973.1 | ||||
XM_017000485.1 | Intron | XP_016855974.1 |