Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC160 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC160 - coiled-coil domain containing 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101357.1 | Intron | NP_001094827.1 | ||||
XM_005262409.4 | Intron | XP_005262466.1 | ||||
XM_005262410.4 | Intron | XP_005262467.1 |