Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607975 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NECAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NECAB2 - N-terminal EF-hand calcium binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019065.2 | 597 | Intron | NP_061938.2 | |||
XM_011523135.1 | 597 | Intron | XP_011521437.1 | |||
XM_017023317.1 | 597 | UTR 5 | XP_016878806.1 |
OSGIN1 - oxidative stress induced growth inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |