Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF20L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF20L1 - PHD finger protein 20-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277196.1 | Intron | NP_001264125.1 | ||||
NM_016018.4 | Intron | NP_057102.4 | ||||
NM_198513.1 | Intron | NP_940915.1 | ||||
XM_005250931.1 | Intron | XP_005250988.1 | ||||
XM_011517075.1 | Intron | XP_011515377.1 | ||||
XM_011517076.1 | Intron | XP_011515378.1 | ||||
XM_011517077.1 | Intron | XP_011515379.1 | ||||
XM_011517078.1 | Intron | XP_011515380.1 | ||||
XM_011517079.1 | Intron | XP_011515381.1 | ||||
XM_011517080.1 | Intron | XP_011515382.1 | ||||
XM_011517081.1 | Intron | XP_011515383.1 | ||||
XM_011517088.1 | Intron | XP_011515390.1 | ||||
XM_017013509.1 | Intron | XP_016868998.1 | ||||
XM_017013510.1 | Intron | XP_016868999.1 | ||||
XM_017013511.1 | Intron | XP_016869000.1 | ||||
XM_017013512.1 | Intron | XP_016869001.1 | ||||
XM_017013513.1 | Intron | XP_016869002.1 | ||||
XM_017013514.1 | Intron | XP_016869003.1 | ||||
XM_017013515.1 | Intron | XP_016869004.1 | ||||
XM_017013516.1 | Intron | XP_016869005.1 | ||||
XM_017013517.1 | Intron | XP_016869006.1 |