Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616786 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPKBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPKBP1 - mitogen-activated protein kinase binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128608.1 | Intron | NP_001122080.1 | ||||
NM_001265611.1 | Intron | NP_001252540.1 | ||||
NM_014994.2 | Intron | NP_055809.2 | ||||
XM_006720438.1 | Intron | XP_006720501.1 | ||||
XM_006720439.2 | Intron | XP_006720502.1 | ||||
XM_011521383.1 | Intron | XP_011519685.1 | ||||
XM_011521384.2 | Intron | XP_011519686.1 | ||||
XM_017022017.1 | Intron | XP_016877506.1 |