Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMD12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAMD12 - sterile alpha motif domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101676.1 | Intron | NP_001095146.1 | ||||
NM_207506.2 | Intron | NP_997389.2 | ||||
XM_011517031.2 | Intron | XP_011515333.1 | ||||
XM_011517034.2 | Intron | XP_011515336.1 | ||||
XM_011517035.2 | Intron | XP_011515337.1 | ||||
XM_017013409.1 | Intron | XP_016868898.1 | ||||
XM_017013410.1 | Intron | XP_016868899.1 | ||||
XM_017013411.1 | Intron | XP_016868900.1 | ||||
XM_017013412.1 | Intron | XP_016868901.1 |