Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
84 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602715 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IPO9 - importin 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMOD1 - leiomodin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHISA4 - shisa family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198149.2 | 534 | Missense Mutation | ATA,ATG | I,M 159 | NP_937792.2 | |
XM_017000419.1 | 534 | Missense Mutation | ATA,ATG | I,M 159 | XP_016855908.1 |
Set Membership: |
HapMap JSNP Validated |