Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612452 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANSL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KANSL1 - KAT8 regulatory NSL complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193465.1 | Intron | NP_001180394.1 | ||||
NM_001193466.1 | Intron | NP_001180395.1 | ||||
NM_015443.3 | Intron | NP_056258.1 | ||||
XM_006721823.1 | Intron | XP_006721886.1 | ||||
XM_006721824.3 | Intron | XP_006721887.1 | ||||
XM_011524628.2 | Intron | XP_011522930.1 | ||||
XM_011524629.2 | Intron | XP_011522931.1 | ||||
XM_011524630.2 | Intron | XP_011522932.1 | ||||
XM_011524631.2 | Intron | XP_011522933.1 | ||||
XM_011524632.2 | Intron | XP_011522934.1 | ||||
XM_017024488.1 | Intron | XP_016879977.1 | ||||
XM_017024489.1 | Intron | XP_016879978.1 |