Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607527 | ||||||||||||||||||||
Literature Links: |
PTBP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTBP3 - polypyrimidine tract binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163788.2 | 3183 | UTR 3 | NP_001157260.1 | |||
NM_001163790.2 | 3183 | UTR 3 | NP_001157262.1 | |||
NM_001244896.1 | 3183 | UTR 3 | NP_001231825.1 | |||
NM_001244897.1 | 3183 | Intron | NP_001231826.1 | |||
NM_001244898.1 | 3183 | UTR 3 | NP_001231827.1 | |||
NM_005156.6 | 3183 | UTR 3 | NP_005147.3 | |||
XM_006717346.1 | 3183 | UTR 3 | XP_006717409.1 | |||
XM_011519265.1 | 3183 | UTR 3 | XP_011517567.1 | |||
XM_017015365.1 | 3183 | UTR 3 | XP_016870854.1 |