Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606025 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR568 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR568 - microRNA 568 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB20 - zinc finger and BTB domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164342.2 | 21010 | UTR 3 | NP_001157814.1 | |||
NM_001164343.2 | 21010 | UTR 3 | NP_001157815.1 | |||
NM_001164344.2 | 21010 | UTR 3 | NP_001157816.1 | |||
NM_001164345.2 | 21010 | UTR 3 | NP_001157817.1 | |||
NM_001164346.2 | 21010 | UTR 3 | NP_001157818.1 | |||
NM_001164347.2 | 21010 | UTR 3 | NP_001157819.1 | |||
NM_015642.5 | 21010 | UTR 3 | NP_056457.3 |
Set Membership: |
HapMap |