Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 117143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BDH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BDH2 - 3-hydroxybutyrate dehydrogenase, type 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CENPE - centromere protein E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286734.1 | Intron | NP_001273663.1 | ||||
NM_001813.2 | Intron | NP_001804.2 | ||||
XM_011531544.2 | Intron | XP_011529846.1 | ||||
XM_011531545.2 | Intron | XP_011529847.1 | ||||
XM_011531546.2 | Intron | XP_011529848.1 | ||||
XM_011531547.2 | Intron | XP_011529849.1 | ||||
XM_011531548.2 | Intron | XP_011529850.1 | ||||
XM_011531549.2 | Intron | XP_011529851.1 | ||||
XM_017007659.1 | Intron | XP_016863148.1 |