Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612896 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC37A8P PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRRC37A8P - leucine rich repeat containing 37 member A8, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDM1 - RAD52 motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034836.1 | Intron | NP_001030008.1 | ||||
NM_001163120.1 | Intron | NP_001156592.1 | ||||
NM_001163121.1 | Intron | NP_001156593.1 | ||||
NM_001163122.1 | Intron | NP_001156594.1 | ||||
NM_001163124.1 | Intron | NP_001156596.1 | ||||
NM_001163125.1 | Intron | NP_001156597.1 | ||||
NM_001163130.1 | Intron | NP_001156602.1 | ||||
NM_145654.3 | Intron | NP_663629.1 | ||||
XM_011524509.2 | Intron | XP_011522811.1 | ||||
XM_011524510.2 | Intron | XP_011522812.1 |
Set Membership: |
HapMap |