Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609199 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAMTSL3 - ADAMTS like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301110.1 | Intron | NP_001288039.1 | ||||
NM_207517.2 | Intron | NP_997400.2 | ||||
XM_011521822.2 | Intron | XP_011520124.1 | ||||
XM_011521823.2 | Intron | XP_011520125.1 | ||||
XM_011521824.2 | Intron | XP_011520126.1 | ||||
XM_011521825.2 | Intron | XP_011520127.1 | ||||
XM_017022434.1 | Intron | XP_016877923.1 | ||||
XM_017022435.1 | Intron | XP_016877924.1 |
Set Membership: |
HapMap Validated |