Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 107580 | |||||||||||||||||||||||
Literature Links: |
TFAP2A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TFAP2A - transcription factor AP-2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032280.2 | 2397 | UTR 3 | NP_001027451.1 | |||
NM_001042425.1 | 2397 | UTR 3 | NP_001035890.1 | |||
NM_003220.2 | 2397 | UTR 3 | NP_003211.1 | |||
XM_006715175.2 | 2397 | UTR 3 | XP_006715238.1 | |||
XM_011514833.2 | 2397 | UTR 3 | XP_011513135.1 | |||
XM_017011232.1 | 2397 | UTR 3 | XP_016866721.1 |
Set Membership: |
HapMap |