Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611343 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDNK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IDNK - IDNK, gluconokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001551.3 | 2502 | Intron | NP_001001551.2 | |||
NM_001256915.1 | 2502 | Intron | NP_001243844.1 | |||
XM_006717110.3 | 2502 | Intron | XP_006717173.1 | |||
XM_006717111.3 | 2502 | UTR 5 | XP_006717174.1 | |||
XM_017014719.1 | 2502 | Intron | XP_016870208.1 | |||
XM_017014720.1 | 2502 | Intron | XP_016870209.1 | |||
XM_017014721.1 | 2502 | Intron | XP_016870210.1 | |||
XM_017014722.1 | 2502 | Intron | XP_016870211.1 | |||
XM_017014723.1 | 2502 | Intron | XP_016870212.1 | |||
XM_017014724.1 | 2502 | UTR 5 | XP_016870213.1 | |||
XM_017014725.1 | 2502 | Intron | XP_016870214.1 |
Set Membership: |
HapMap |