Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608313 MIM: 605042 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARG1 - arginase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000045.3 | Intron | NP_000036.2 | ||||
NM_001244438.1 | Intron | NP_001231367.1 | ||||
XM_011535801.2 | Intron | XP_011534103.1 |
MED23 - mediator complex subunit 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270521.1 | Intron | NP_001257450.1 | ||||
NM_001270522.1 | Intron | NP_001257451.1 | ||||
NM_004830.3 | Intron | NP_004821.2 | ||||
NM_015979.3 | Intron | NP_057063.2 | ||||
XM_005267223.2 | Intron | XP_005267280.1 | ||||
XM_006715612.2 | Intron | XP_006715675.1 | ||||
XM_011536257.2 | Intron | XP_011534559.1 | ||||
XM_017011501.1 | Intron | XP_016866990.1 |
Set Membership: |
HapMap |