Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTCCAGGGAACCGAACTCAGTT[A/G]CAGCACTCTTCACCTTGTAATGGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609448 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTF3L4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTF3L4 - basic transcription factor 3 like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136497.2 | Intron | NP_001129969.1 | ||||
NM_001243767.1 | Intron | NP_001230696.1 | ||||
NM_152265.4 | Intron | NP_689478.1 |
TXNDC12 - thioredoxin domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015913.3 | Intron | NP_056997.1 |
TXNDC12-AS1 - TXNDC12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |