Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
MIR4273 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MIR4273 - microRNA 4273 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF717 - zinc finger protein 717 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128223.2 | Intron | NP_001121695.1 | ||||
NM_001290208.1 | Intron | NP_001277137.1 | ||||
NM_001290209.1 | Intron | NP_001277138.1 | ||||
NM_001290210.1 | Intron | NP_001277139.1 | ||||
NM_001324026.1 | Intron | NP_001310955.1 | ||||
NM_001324027.1 | Intron | NP_001310956.1 | ||||
NM_001324028.1 | Intron | NP_001310957.1 | ||||
XM_005264711.3 | Intron | XP_005264768.1 | ||||
XM_006712912.3 | Intron | XP_006712975.1 | ||||
XM_011533244.2 | Intron | XP_011531546.1 | ||||
XM_011533245.2 | Intron | XP_011531547.1 | ||||
XM_011533246.2 | Intron | XP_011531548.1 | ||||
XM_011533248.2 | Intron | XP_011531550.1 | ||||
XM_011533249.2 | Intron | XP_011531551.1 | ||||
XM_011533250.2 | Intron | XP_011531552.1 | ||||
XM_011533251.1 | Intron | XP_011531553.1 | ||||
XM_017005486.1 | Intron | XP_016860975.1 | ||||
XM_017005487.1 | Intron | XP_016860976.1 |
Set Membership: |
HapMap |