Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601998 MIM: 605720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRRA - estrogen related receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR137 - G protein-coupled receptor 137 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK4 - potassium two pore domain channel subfamily K member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317090.1 | 678 | Silent Mutation | CCA,CCG | P,P 164 | NP_001304019.1 | |
NM_033310.2 | 678 | Silent Mutation | CCA,CCG | P,P 164 | NP_201567.1 |
KCNK4-TEX40 - KCNK4-TEX40 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX40 - testis expressed 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |