Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601837 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD13 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD13 - abhydrolase domain containing 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIG4 - DNA ligase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098268.1 | 3375 | UTR 3 | NP_001091738.1 | |||
NM_002312.3 | 3375 | UTR 3 | NP_002303.2 | |||
NM_206937.1 | 3375 | UTR 3 | NP_996820.1 | |||
XM_005254056.1 | 3375 | UTR 3 | XP_005254113.1 | |||
XM_005254057.4 | 3375 | UTR 3 | XP_005254114.1 | |||
XM_005254058.3 | 3375 | UTR 3 | XP_005254115.1 | |||
XM_006719951.3 | 3375 | UTR 3 | XP_006720014.1 | |||
XM_006719952.1 | 3375 | UTR 3 | XP_006720015.1 | |||
XM_011521091.2 | 3375 | UTR 3 | XP_011519393.1 | |||
XM_011521092.2 | 3375 | UTR 3 | XP_011519394.1 | |||
XM_017020563.1 | 3375 | UTR 3 | XP_016876052.1 | |||
XM_017020564.1 | 3375 | UTR 3 | XP_016876053.1 | |||
XM_017020565.1 | 3375 | UTR 3 | XP_016876054.1 | |||
XM_017020566.1 | 3375 | UTR 3 | XP_016876055.1 | |||
XM_017020567.1 | 3375 | UTR 3 | XP_016876056.1 | |||
XM_017020568.1 | 3375 | UTR 3 | XP_016876057.1 | |||
XM_017020569.1 | 3375 | UTR 3 | XP_016876058.1 | |||
XM_017020570.1 | 3375 | UTR 3 | XP_016876059.1 | |||
XM_017020571.1 | 3375 | UTR 3 | XP_016876060.1 | |||
XM_017020572.1 | 3375 | UTR 3 | XP_016876061.1 | |||
XM_017020573.1 | 3375 | UTR 3 | XP_016876062.1 |
Set Membership: |
HapMap |