Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606475 MIM: 602139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD320 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD320 - CD320 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165895.1 | 906 | Silent Mutation | TCA,TCG | S,S 238 | NP_001159367.1 | |
NM_016579.3 | 906 | Silent Mutation | TCA,TCG | S,S 280 | NP_057663.1 |
NDUFA7 - NADH:ubiquinone oxidoreductase subunit A7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |