Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611782 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSCAML1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DSCAML1 - DS cell adhesion molecule like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020693.3 | 4650 | UTR 3 | NP_065744.2 | |||
XM_011542917.1 | 4650 | UTR 3 | XP_011541219.1 | |||
XM_011542918.1 | 4650 | UTR 3 | XP_011541220.1 | |||
XM_011542919.2 | 4650 | UTR 3 | XP_011541221.1 | |||
XM_011542920.2 | 4650 | UTR 3 | XP_011541222.1 | |||
XM_011542921.2 | 4650 | UTR 3 | XP_011541223.1 | |||
XM_011542924.2 | 4650 | UTR 3 | XP_011541226.1 | |||
XM_011542925.2 | 4650 | UTR 3 | XP_011541227.1 |
Set Membership: |
HapMap |