Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605874 | ||||||||||||||||||||
Literature Links: |
KCNK9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNK9 - potassium two pore domain channel subfamily K member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282534.1 | Intron | NP_001269463.1 | ||||
XM_011517101.2 | Intron | XP_011515403.1 | ||||
XM_011517102.2 | Intron | XP_011515404.1 | ||||
XM_011517103.2 | Intron | XP_011515405.1 | ||||
XM_017013530.1 | Intron | XP_016869019.1 | ||||
XM_017013531.1 | Intron | XP_016869020.1 |