Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603897 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MATN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MATN4 - matrilin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003833.4 | 1987 | Missense Mutation | AAA,AAC | K,N 575 | NP_003824.2 | |
NM_030590.3 | 1987 | Missense Mutation | AAA,AAC | K,N 534 | NP_085080.1 | |
NM_030592.3 | 1987 | Missense Mutation | AAA,AAC | K,N 493 | NP_085095.1 | |
XM_005260597.1 | 1987 | Missense Mutation | AAA,AAC | K,N 575 | XP_005260654.1 | |
XM_017028113.1 | 1987 | UTR 3 | XP_016883602.1 | |||
XM_017028114.1 | 1987 | UTR 3 | XP_016883603.1 | |||
XM_017028115.1 | 1987 | UTR 3 | XP_016883604.1 |
Set Membership: |
HapMap |