Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MINPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MINPP1 - multiple inositol-polyphosphate phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178117.1 | 1773 | UTR 5 | NP_001171588.1 | |||
NM_001178118.1 | 1773 | Intron | NP_001171589.1 | |||
NM_004897.4 | 1773 | UTR 5 | NP_004888.2 | |||
XM_006718078.2 | 1773 | UTR 5 | XP_006718141.1 | |||
XM_011540379.2 | 1773 | Intron | XP_011538681.1 | |||
XM_017016965.1 | 1773 | UTR 5 | XP_016872454.1 | |||
XM_017016966.1 | 1773 | Intron | XP_016872455.1 |
MIR4678 - microRNA 4678 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |