Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608750 MIM: 608721 MIM: 610145 MIM: 611620 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALG3 - ALG3, alpha-1,3- mannosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CAMK2N2 - calcium/calmodulin dependent protein kinase II inhibitor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ECE2 - endothelin converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1224 - microRNA 1224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA5B2 - von Willebrand factor A domain containing 5B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |