Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607068 MIM: 607072 MIM: 607069 MIM: 607070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB561D2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYB561D2 - cytochrome b561 family member D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291284.1 | 104 | UTR 5 | NP_001278213.1 | |||
NM_007022.4 | 104 | UTR 5 | NP_008953.1 |
NPRL2 - NPR2-like, GATOR1 complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006545.4 | 104 | UTR 5 | NP_006536.3 | |||
XM_005264808.4 | 104 | Intron | XP_005264865.1 | |||
XM_011533288.2 | 104 | Intron | XP_011531590.1 | |||
XM_017005555.1 | 104 | Intron | XP_016861044.1 | |||
XM_017005556.1 | 104 | Intron | XP_016861045.1 |
TMEM115 - transmembrane protein 115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYND10 - zinc finger MYND-type containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |