Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCP11 - t-complex 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093728.2 | 1538 | Silent Mutation | CTA,CTG | L,L 421 | NP_001087197.1 | |
NM_001261817.1 | 1538 | Silent Mutation | CTA,CTG | L,L 416 | NP_001248746.1 | |
NM_001261818.1 | 1538 | Silent Mutation | CTA,CTG | L,L 375 | NP_001248747.1 | |
NM_001261819.1 | 1538 | Silent Mutation | CTA,CTG | L,L 370 | NP_001248748.1 | |
NM_001261820.1 | 1538 | Silent Mutation | CTA,CTG | L,L 345 | NP_001248749.1 | |
NM_001261821.1 | 1538 | Silent Mutation | CTA,CTG | L,L 345 | NP_001248750.1 | |
NM_018679.5 | 1538 | Silent Mutation | CTA,CTG | L,L 346 | NP_061149.1 | |
XM_005249339.2 | 1538 | Silent Mutation | CTA,CTG | L,L 267 | XP_005249396.1 | |
XM_011514830.1 | 1538 | Silent Mutation | CTA,CTG | L,L 524 | XP_011513132.1 | |
XM_011514831.1 | 1538 | Silent Mutation | CTA,CTG | L,L 267 | XP_011513133.1 | |
XM_011514832.1 | 1538 | Silent Mutation | CTA,CTG | L,L 267 | XP_011513134.1 |