Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300697 | ||||||||||||||||||||
Literature Links: |
HUWE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HUWE1 - HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031407.6 | Intron | NP_113584.3 | ||||
XM_005261965.3 | Intron | XP_005262022.1 | ||||
XM_011530747.1 | Intron | XP_011529049.1 | ||||
XM_011530751.2 | Intron | XP_011529053.1 | ||||
XM_017029191.1 | Intron | XP_016884680.1 | ||||
XM_017029192.1 | Intron | XP_016884681.1 | ||||
XM_017029193.1 | Intron | XP_016884682.1 | ||||
XM_017029194.1 | Intron | XP_016884683.1 | ||||
XM_017029195.1 | Intron | XP_016884684.1 | ||||
XM_017029196.1 | Intron | XP_016884685.1 | ||||
XM_017029197.1 | Intron | XP_016884686.1 | ||||
XM_017029198.1 | Intron | XP_016884687.1 | ||||
XM_017029199.1 | Intron | XP_016884688.1 | ||||
XM_017029200.1 | Intron | XP_016884689.1 | ||||
XM_017029201.1 | Intron | XP_016884690.1 | ||||
XM_017029202.1 | Intron | XP_016884691.1 | ||||
XM_017029203.1 | Intron | XP_016884692.1 | ||||
XM_017029204.1 | Intron | XP_016884693.1 | ||||
XM_017029205.1 | Intron | XP_016884694.1 | ||||
XM_017029206.1 | Intron | XP_016884695.1 |