Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609911 MIM: 191050 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A47 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC25A47 - solute carrier family 25 member 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004184.3 | Intron | NP_004175.2 | ||||
NM_173701.1 | Intron | NP_776049.1 | ||||
NM_213645.1 | Intron | NP_998810.1 | ||||
NM_213646.1 | Intron | NP_998811.1 | ||||
XM_005268044.3 | Intron | XP_005268101.1 | ||||
XM_006720249.3 | Intron | XP_006720312.1 | ||||
XM_011537133.2 | Intron | XP_011535435.1 | ||||
XM_011537135.2 | Intron | XP_011535437.1 | ||||
XM_011537136.2 | Intron | XP_011535438.1 | ||||
XM_017021627.1 | Intron | XP_016877116.1 | ||||
XM_017021628.1 | Intron | XP_016877117.1 | ||||
XM_017021629.1 | Intron | XP_016877118.1 |
Set Membership: |
HapMap |