Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612062 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf31 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C22orf31 - chromosome 22 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015370.1 | 747 | Silent Mutation | CAC,CAT | H,H 275 | NP_056185.1 | |
XM_005261491.3 | 747 | Silent Mutation | CAC,CAT | H,H 171 | XP_005261548.1 | |
XM_011530096.2 | 747 | Silent Mutation | CAC,CAT | H,H 234 | XP_011528398.1 | |
XM_017028741.1 | 747 | Silent Mutation | CAC,CAT | H,H 234 | XP_016884230.1 |
ZNRF3 - zinc and ring finger 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |