Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAACCTTCCAAAGAAGTGCCTTC[A/G]TTTATTTACAGGGCTTAGGAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608955 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506679 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100506679 - uncharacterized LOC100506679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL1 - tubulin tyrosine ligase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012263.4 | 1654 | UTR 3 | NP_036395.1 | |||
XM_006724225.3 | 1654 | UTR 3 | XP_006724288.1 | |||
XM_011530108.2 | 1654 | UTR 3 | XP_011528410.1 | |||
XM_011530110.2 | 1654 | UTR 3 | XP_011528412.1 | |||
XM_017028751.1 | 1654 | UTR 3 | XP_016884240.1 | |||
XM_017028752.1 | 1654 | UTR 3 | XP_016884241.1 | |||
XM_017028753.1 | 1654 | UTR 3 | XP_016884242.1 |
Set Membership: |
HapMap |