Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300187 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRPX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SRPX - sushi repeat containing protein, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170750.1 | 1283 | Intron | NP_001164221.1 | |||
NM_001170751.1 | 1283 | Intron | NP_001164222.1 | |||
NM_001170752.1 | 1283 | Intron | NP_001164223.1 | |||
NM_006307.4 | 1283 | Intron | NP_006298.1 | |||
XM_017029893.1 | 1283 | Silent Mutation | AGC,AGT | S,S 404 | XP_016885382.1 |