Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604782 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASH2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ASH2L - ASH2 like histone lysine methyltransferase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105214.2 | 757 | Intron | NP_001098684.1 | |||
NM_001261832.1 | 757 | Intron | NP_001248761.1 | |||
NM_001282272.1 | 757 | UTR 5 | NP_001269201.1 | |||
NM_004674.4 | 757 | Silent Mutation | TCC,TCT | S,S 57 | NP_004665.2 | |
XM_005273682.1 | 757 | Silent Mutation | TCC,TCT | S,S 57 | XP_005273739.1 | |
XM_005273683.1 | 757 | Intron | XP_005273740.1 | |||
XM_006716412.1 | 757 | Silent Mutation | TCC,TCT | S,S 57 | XP_006716475.1 | |
XM_006716413.2 | 757 | Intron | XP_006716476.1 |
Set Membership: |
HapMap Validated |