Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601440 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf67 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C17orf67 - chromosome 17 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085430.2 | Intron | NP_001078899.2 | ||||
XM_011524735.2 | Intron | XP_011523037.1 |
DGKE - diacylglycerol kinase epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003647.2 | Intron | NP_003638.1 | ||||
XM_011525394.2 | Intron | XP_011523696.1 | ||||
XM_011525395.2 | Intron | XP_011523697.1 | ||||
XM_011525396.2 | Intron | XP_011523698.1 | ||||
XM_017025243.1 | Intron | XP_016880732.1 | ||||
XM_017025244.1 | Intron | XP_016880733.1 | ||||
XM_017025245.1 | Intron | XP_016880734.1 |
Set Membership: |
HapMap |