Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTAGCTGTGGCGAGTTATTCTAC[C/T]GTGCACTAAAGTCGGGAGAAACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612451 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372653 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105372653 - uncharacterized LOC105372653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF114 - ring finger protein 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018683.3 | Intron | NP_061153.1 |