Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604770 MIM: 606540 MIM: 615645 | ||||||||||||||||||||
Literature Links: |
ACAA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAA2 - acetyl-CoA acyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006111.2 | Intron | NP_006102.2 |
MYO5B - myosin VB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA17 - small Cajal body-specific RNA 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG22 - small nucleolar RNA host gene 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |