Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191840 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf55 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf55 - chromosome 10 open reading frame 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001791.2 | 2297 | UTR 3 | NP_001001791.2 |
PLAU - plasminogen activator, urokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145031.2 | 2297 | Intron | NP_001138503.1 | |||
NM_001319191.1 | 2297 | Intron | NP_001306120.1 | |||
NM_002658.4 | 2297 | Intron | NP_002649.1 | |||
XM_011539866.2 | 2297 | Intron | XP_011538168.1 |
Set Membership: |
HapMap |