Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG16L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATG16L2 - autophagy related 16 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318766.1 | Intron | NP_001305695.1 | ||||
NM_033388.1 | Intron | NP_203746.1 | ||||
XM_005274376.4 | Intron | XP_005274433.1 | ||||
XM_006718732.2 | Intron | XP_006718795.1 | ||||
XM_006718733.3 | Intron | XP_006718796.1 | ||||
XM_006718734.2 | Intron | XP_006718797.1 | ||||
XM_011545332.1 | Intron | XP_011543634.1 | ||||
XM_011545333.1 | Intron | XP_011543635.1 | ||||
XM_011545334.1 | Intron | XP_011543636.1 |