Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf106 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf106 - chromosome 1 open reading frame 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142569.2 | 385 | Silent Mutation | AGC,AGT | S,S 8 | NP_001136041.1 | |
NM_018265.3 | 385 | Silent Mutation | AGC,AGT | S,S 107 | NP_060735.3 | |
XM_011509754.2 | 385 | Silent Mutation | AGC,AGT | S,S 8 | XP_011508056.1 | |
XM_011509755.1 | 385 | Silent Mutation | AGC,AGT | S,S 8 | XP_011508057.1 | |
XM_011509756.2 | 385 | Silent Mutation | AGC,AGT | S,S 107 | XP_011508058.1 |