Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HACL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HACL1 - 2-hydroxyacyl-CoA lyase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284413.1 | 1730 | Silent Mutation | CAA,CAG | Q,Q 420 | NP_001271342.1 | |
NM_001284415.1 | 1730 | Silent Mutation | CAA,CAG | Q,Q 387 | NP_001271344.1 | |
NM_001284416.1 | 1730 | Silent Mutation | CAA,CAG | Q,Q 365 | NP_001271345.1 | |
NM_012260.3 | 1730 | Silent Mutation | CAA,CAG | Q,Q 447 | NP_036392.2 |