Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604939 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2R1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLA2R1 - phospholipase A2 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007267.2 | 4851 | Intron | NP_001007268.1 | |||
NM_001195641.1 | 4851 | Intron | NP_001182570.1 | |||
NM_007366.4 | 4851 | Intron | NP_031392.3 | |||
XM_005246392.3 | 4851 | UTR 3 | XP_005246449.1 | |||
XM_011510820.2 | 4851 | UTR 3 | XP_011509122.1 | |||
XM_017003598.1 | 4851 | Intron | XP_016859087.1 | |||
XM_017003599.1 | 4851 | Intron | XP_016859088.1 | |||
XM_017003600.1 | 4851 | Intron | XP_016859089.1 | |||
XM_017003601.1 | 4851 | Intron | XP_016859090.1 | |||
XM_017003602.1 | 4851 | Intron | XP_016859091.1 |