Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601625 MIM: 610625 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ART1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ART1 - ADP-ribosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ART5 - ADP-ribosyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079536.1 | 1452 | UTR 3 | NP_001073004.1 | |||
NM_001297668.1 | 1452 | UTR 3 | NP_001284597.1 | |||
NM_053017.4 | 1452 | UTR 3 | NP_443750.2 | |||
XM_005252779.3 | 1452 | UTR 3 | XP_005252836.1 | |||
XM_006718136.3 | 1452 | UTR 3 | XP_006718199.1 | |||
XM_011519879.2 | 1452 | UTR 3 | XP_011518181.1 | |||
XM_017017171.1 | 1452 | UTR 3 | XP_016872660.1 |
TRPC2 - transient receptor potential cation channel subfamily C member 2, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |