Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614663 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALY PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RALY - RALY heterogeneous nuclear ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007367.3 | Intron | NP_031393.2 | ||||
NM_016732.2 | Intron | NP_057951.1 | ||||
XM_005260334.4 | Intron | XP_005260391.1 | ||||
XM_005260336.4 | Intron | XP_005260393.1 | ||||
XM_011528694.2 | Intron | XP_011526996.1 | ||||
XM_011528695.2 | Intron | XP_011526997.1 | ||||
XM_017027731.1 | Intron | XP_016883220.1 |
RALY-AS1 - RALY antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |